Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM46A Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299201AF594
Description
FAM46A Polyclonal specifically detects FAM46A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
FAM46A | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
C6orf37, chromosome 6 open reading frame 37, family with sequence similarity 46, member A, FLJ20037, FLJ31495, HBV XAg-transactivated protein 11, HBV X-transactivated gene 11 protein, hypothetical protein LOC55603, retinal expressed gene C6orf37, XTP11 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human FAM46A. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
55603 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction