Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM46A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120184
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120184 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120184 Supplier Invitrogen™ Supplier No. PA5120184
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HT-29, A-431, SW480, SW620, A375, Mouse eye, Mouse small intestine. Immunogen sequence: PLTLKEAYVQ KMVKVCNDSD RWSLISLSNN SGKNVELKFV DSLRRQFEFS VDSFQIKLDS LLLFYECSEN PMTETFHPTI IGESVYGDFQ EAFDHLCNKI IATRNPEEIR GGGLLKYCNL LVRGFRPASD EIKTLQRYMC SRFFIDFSDI GEQQRKLESY LQNHFVGLED RKYEYLMTLH GVVNESTVCL MGHERRQTLN LITMLAIRVL ADQNVIPNVA N.

Cytoplasmic non-canonical poly(A) RNA polymerase that catalyzes the transfer of one adenosine molecule from an ATP to an mRNA poly(A) tail bearing a 3'-OH terminal group and participates in the cytoplasmic polyadenylation. Polyadenylates mRNA encoding extracellular matrix constituents and other genes crucial for bone mineralization and during osteoblast mineralization, mainly focuses on ER-targeted mRNAs (By similarity).
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM46A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.44 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene FAM46A
Gene Accession No. Q96IP4
Gene Alias C6orf37; FAM46A; family with sequence similarity 46 member A; family with sequence similarity 46, member A; HBV XAg-transactivated protein 11; HBV X-transactivated gene 11 protein; protein FAM46A; retinal expressed gene C6orf37; TENT5A; Terminal nucleotidyltransferase 5A; XTP11
Gene Symbols FAM46A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 180-400 of human FAM46A (NP_060103.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 212943, 55603
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.