Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM46B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113297
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113297 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113297 Supplier Invitrogen™ Supplier No. PA5113297
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM46B Polyclonal Antibody for Western Blot, ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM46B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene TENT5B
Gene Accession No. Q96A09
Gene Alias FAM46B; family with sequence similarity 46 member B; family with sequence similarity 46, member B; protein FAM46B; TENT5B; Terminal nucleotidyltransferase 5B
Gene Symbols TENT5B
Host Species Rabbit
Immunogen Recombinant Human Terminal nucleotidyltransferase 5B protein (2-425AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115572
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.