Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM46C Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | FAM46C |
|---|---|
| Concentration | 0.3 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-4686
![]()
|
Proteintech Group Inc
250381AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
FAM46C belongs to the FAM46 family. The exact function of FAM46C remains unknown.Specifications
| FAM46C | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q5VWP2 | |
| 54855 | |
| FAM46C Fusion Protein Ag19147 | |
| Primary | |
| -20°C | |
| FAM46C |
| 0.3 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| FAM46C, Protein FAM46C | |
| FAM46C | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title