Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM46D Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP216417
Description
FAM46D Polyclonal specifically detects FAM46D in Mouse samples. It is validated for Western Blot.Specifications
FAM46D | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
cancer/testis antigen 112, CT1.26, CT112, family with sequence similarity 46, member D, hypothetical protein LOC169966, MGC26999 | |
Rabbit | |
45 kDa | |
0.1 mL | |
Primary | |
Mouse. Expected to cross react based on sequence identity: Human (100%), Monkey (95%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot | |
1mg/mL | |
Western Blot 1:1000-1:10000 | |
Q8NEK8 | |
FAM46D | |
Recombinant protein encompassing a sequence within the center region of human FAM46D. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
169966 | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction