Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM46D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531591
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531591 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531591 Supplier Invitrogen™ Supplier No. PA531591
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: mouse Liver. Predicted reactivity: Mouse (81%), Rat (81%), Rhesus Monkey (95%), Bovine (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Antibodies against the protein encoded by this gene were found only in plasma from cancer patients. While it may be a target for immunotherapy, the function of this gene is unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM46D
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene FAM46D
Gene Accession No. Q8NEK8
Gene Alias cancer/testis antigen 112; CT1.26; CT112; FAM46D; family with sequence similarity 46 member D; family with sequence similarity 46, member D; Gm377; Non-canonical poly(A) polymerase FAM46D; protein FAM46D; TENT5D; Terminal nucleotidyltransferase 5D
Gene Symbols FAM46D
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human FAM46D. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 169966, 213449
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.