Learn More
Invitrogen™ FAM46D Polyclonal Antibody

Description
Immunogen sequence: PTMEVKPKDI IHVVKDQLIG QGIIVKDARL NGSVASYILA SHNGISYKDL DVIFGVELPG NEEFQV Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 85%.
Specifications
Specifications
| Antigen | FAM46D |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | FAM46D |
| Gene Accession No. | Q8NEK8 |
| Gene Alias | cancer/testis antigen 112; CT1.26; CT112; FAM46D; family with sequence similarity 46 member D; family with sequence similarity 46, member D; Gm377; Non-canonical poly(A) polymerase FAM46D; protein FAM46D; TENT5D; Terminal nucleotidyltransferase 5D |
| Gene Symbols | FAM46D |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.