Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM46D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569909
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569909 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569909 Supplier Invitrogen™ Supplier No. PA569909
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Human: 100%; Pig: 100%.

Antibodies against the protein encoded by this gene were found only in plasma from cancer patients. While it may be a target for immunotherapy, the function of this gene is unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM46D
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FAM46D
Gene Accession No. Q8NEK8
Gene Alias cancer/testis antigen 112; CT1.26; CT112; FAM46D; family with sequence similarity 46 member D; family with sequence similarity 46, member D; Gm377; Non-canonical poly(A) polymerase FAM46D; protein FAM46D; TENT5D; Terminal nucleotidyltransferase 5D
Gene Symbols FAM46D
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human FAM46D.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 169966
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.