Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM47B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562285
Description
Immunogen sequence: ARARCEGQEM TTEELTKPGK YHFWESCPRP FESRMPHLRL VLPITRRMAS LCLKPPKTRR Highest antigen sequence identity to the following orthologs: Mouse - 31%, Rat - 30%.
FAM47B is a protein coding gene.
Specifications
FAM47B | |
Polyclonal | |
Unconjugated | |
FAM47B | |
FAM47B; family with sequence similarity 47 member B; family with sequence similarity 47, member B; protein FAM47B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
170062 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NA70 | |
FAM47B | |
Recombinant protein corresponding to Human FAM47B. Recombinant protein control fragment (Product #RP-100084). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction