Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM47E Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546681
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546681 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546681 Supplier Invitrogen™ Supplier No. PA546681
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: KLEDTWAYCQ DTRKGMKEPT KLLKKHSTQV YLGPSKKTSV SNAGQWLYEE Sequence homology: Human: 100%; Mouse: 85%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM47E
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FAM47E
Gene Accession No. Q6ZV65
Gene Alias C730043O17; FAM47E; family with sequence similarity 47 member E; family with sequence similarity 47, member E; Gm1381; Protein FAM47E
Gene Symbols FAM47E
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human FAM47E.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100129583
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.