Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM48A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558586
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558586 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558586 Supplier Invitrogen™ Supplier No. PA558586
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DSETIRLPYE EGELLEYLDA EELPPILVDL LEKSQVNIFH CGCVIAEIRD YRQSSNMKSP GYQSRHILLR PTMQTLICDV HSITSDNHKW TQED Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

Required for MAP kinase p38 (MAPK11, MAPK12, MAPK13 and/or MAPK14) activation during gastrulation. Required for down-regulation of E-cadherin during gastrulation by regulating E-cadherin protein level downstream from NCK-interacting kinase (NIK) and independently of the regulation of transcription by FGF signaling and Snail. Required for starvation-induced ATG9A trafficking during autophagy. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM48A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SUPT20H
Gene Accession No. Q8NEM7
Gene Alias bA421P11.4; C13; C13orf19; Fam48a; family with sequence similarity 48, member A; FP757; p38-interacting protein; P38IP; protein FAM48A; RGD1307812; SPT20; SPT20 homolog, SAGA complex component; suppressor of Ty 20; suppressor of Ty 20 homolog; Supt20; SUPT20H; transcription factor (p38 interacting protein); transcription factor SPT20 homolog; tumor rejection antigen
Gene Symbols SUPT20H
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM48A. Recombinant protein control fragment (Product #RP-97915).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55578
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.