Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM48A Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983104
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3104 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3104 Supplier Bioss Supplier No. BS13720R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Required for MAP kinase p38 (MAPK11, MAPK12, MAPK13 and/or MAPK14) activation during gastrulation. Required for down-regulation of E-cadherin during gastrulation by regulating E-cadherin protein level downstream from NCK-interacting kinase (NIK) and independently of the regulation of transcription by Fgf signaling and Snail.

Specifications

Antigen FAM48A
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SUPT20H
Gene Accession No. Q8NEM7
Gene Alias bA421P11.4; C13; C13orf19; FAM48A; family with sequence similarity 48, member A; FP757; p38-interacting protein; p38IP; protein FAM48A; SPT20; SPT20 homolog, SAGA complex component; suppressor of Ty 20 homolog; SUPT20H; transcription factor (p38 interacting protein); Transcription factor SPT20 homolog; tumor rejection antigen
Gene Symbols SUPT20H
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FAM48A.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55578
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.