Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM49A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA525278
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA525278 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA525278 Supplier Invitrogen™ Supplier No. PA525278
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with bovine based on sequence homology.

FAM49A is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM49A
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene FAM49A
Gene Accession No. Q8BHZ0, Q9H0Q0
Gene Alias 2410157M17Rik; 9630047E15; D12Ertd553e; FAM49A; family with sequence similarity 49 member A; family with sequence similarity 49, member A; family with sequence similarity 49, member B; protein FAM49A
Gene Symbols FAM49A
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 5-33 amino acids from the N-terminal region of human FA49A.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 76820, 81553
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.