Learn More
FAM49A Polyclonal Antibody

Description
FAM49A Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | FAM49A |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 |
| Gene | FAM49A |
| Gene Accession No. | Q9H0Q0 |
| Gene Alias | 2410157M17Rik; 9630047E15; D12Ertd553e; FAM49A; family with sequence similarity 49 member A; family with sequence similarity 49, member A; family with sequence similarity 49, member B; protein FAM49A |
| Gene Symbols | FAM49A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.