Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM49B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552647
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552647 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552647 Supplier Invitrogen™ Supplier No. PA552647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GNLLKVLTCT DLEQGPNFFL DFENAQPTES EKEIYNQVNV VLKDAEGILE DLQSYRGAGH EIREAIQHPA DEKLQEKAWG AVVPLVGKLK KFYEFSQRLE AALRGLLGAL TSTPYSPTQH LEREQALAKQ FAE Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

Negatively regulates RAC1 signaling and RAC1-driven cytoskeletal remodeling (PubMed:31285585, PubMed:30250061). Regulates chemotaxis, cell migration and epithelial polarization by controlling the polarity, plasticity, duration and extent of protrusions. Limits Rac1 mediated activation of the Scar/WAVE complex, focuses protrusion signals and regulates pseudopod complexity by inhibiting Scar/WAVE-induced actin polymerization (PubMed:30250061). Protects against Salmonella bacterial infection. Attenuates processes such as macropinocytosis, phagocytosis and cell migration and restrict sopE-mediated bacterial entry (PubMed:31285585). Restricts also infection mediated by Mycobacterium tuberculosis and Listeria monocytogenes. Involved in the regulation of mitochondrial dynamics and oxidative stress (PubMed:29059164). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM49B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CYRIB
Gene Accession No. Q9NUQ9
Gene Alias 0910001A06Rik; AW122079; BM-009; FAM49B; family with sequence similarity 49 member B; family with sequence similarity 49, member B; L1; Protein FAM49B; RGD1307697
Gene Symbols CYRIB
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM49B. Recombinant protein control fragment (Product #RP-89928).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51571
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.