Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM50A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $648.50
Specifications
Antigen | FAM50A |
---|---|
Dilution | Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunoprecipitation, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
FAM50A Polyclonal antibody specifically detects FAM50A in Human, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin).Specifications
FAM50A | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
RUO | |
Human, Zebrafish, Rat | |
DXS9928EXAP-5 protein, family with sequence similarity 50, member A, HXC-26, Protein HXC-26, Protein XAP-5, XAP5HXC26,9F | |
FAM50A | |
IgG | |
Affinity Purified | |
Specificity of human FAM50A antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunoprecipitation, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Polyclonal | |
Rabbit | |
DNA replication Transcription Translation and Splicing | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
9130 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:VTLNDMKAKQEALVKEREKQLAKKEQSKELQMKLEKLREKERKKEAKRKISSLSFTLEEEEEGGEEEEEAAMYEEEMEREEITTKKRKLGKNPDVDTSFLPDRDRE | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title