Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM50B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$499.50
Specifications
| Antigen | FAM50B |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
FAM50B Polyclonal specifically detects FAM50B in Human samples. It is validated for Western Blot.Specifications
| FAM50B | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 26240 | |
| Synthetic peptides corresponding to FAM50B(family with sequence similarity 50, member B) The peptide sequence was selected from the N terminal of FAM50B. Peptide sequence EQRRERKRKISCLSFALDDLDDQADAAEARRAGNLGKNPDVDTSFLPDRD. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| family with sequence similarity 50, member B, Protein XAP-5-like, X5LD6S2654E | |
| FAM50B | |
| IgG | |
| 39 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title