Learn More
Invitrogen™ FAM53C Polyclonal Antibody

Description
Immunogen sequence: SNCGNSFQLV SEGASWRGLP HCSCAEFQDS LNFSYHPSGL SLHLRPPSRG For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 93%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.
Specifications
Specifications
| Antigen | FAM53C |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | FAM53C |
| Gene Accession No. | Q9NYF3 |
| Gene Alias | 2810012G03Rik; C5orf6; FAM53C; family with sequence similarity 53 member C; family with sequence similarity 53, member C; Gm21179; Protein FAM53C; putative nuclear protein |
| Gene Symbols | FAM53C |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.