Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM54A Monoclonal Antibody (OTI1A7)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA527463
Description
FAM54A Monoclonal Antibody for Western Blot, IHC (P)
FAM54A belongs to the MTFR1/FAM54 family. The function of the FAM54A protein is not known.
Specifications
FAM54A | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6P444 | |
MTFR2 | |
Full length human recombinant protein of FAM54A produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1A7 | |
Unconjugated | |
MTFR2 | |
DUF729 domain containing 1; DUF729 domain-containing protein 1; DUFD1; EGK_15321; FAM54A; family with sequence similarity 54, member A; LOC100911069; Mitochondrial fission regulator 2; Mtfr2; protein FAM54A; protein FAM54A-like | |
Mouse | |
Affinity Chromatography | |
RUO | |
113115 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction