Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM54A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556259
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556259 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556259 Supplier Invitrogen™ Supplier No. PA556259
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LILNILREML EYFGVPVEQV LLIWENKDYG STRSIVRIIG KMLPLEPCRR PNFELIPLLN SVDSDNCGSM VPSFADILYV ANDEEASYL Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 67%.

FAM54A belongs to the MTFR1/FAM54 family. The function of the FAM54A protein is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM54A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MTFR2
Gene Accession No. Q6P444
Gene Alias DUF729 domain containing 1; DUF729 domain-containing protein 1; DUFD1; EGK_15321; FAM54A; family with sequence similarity 54, member A; LOC100911069; Mitochondrial fission regulator 2; Mtfr2; protein FAM54A; protein FAM54A-like
Gene Symbols MTFR2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM54A. Recombinant protein control fragment (Product #RP-94896).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113115
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.