Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM55A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113244
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113244 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113244 Supplier Invitrogen™ Supplier No. PA5113244
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM55A Polyclonal Antibody for ELISA, IHC (P), Western Blot

Predicted to be located in extracellular region. Predicted to be integral component of membrane. NXPE1 (Neurexophilin And PC-Esterase Domain Family Member 1) is a Protein Coding gene. An important paralog of this gene is NXPE2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM55A
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene NXPE1
Gene Accession No. Q8N323
Gene Alias FAM55A; family with sequence similarity 55, member A; neurexophilin and PC-esterase domain family member 1; neurexophilin and PC-esterase domain family, member 1; NXPE family member 1; NXPE1; Protein FAM55A
Gene Symbols NXPE1
Host Species Rabbit
Immunogen Recombinant Human NXPE family member 1 protein (1-405AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 120400
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.