Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Invitrogen™ FAM55D Polyclonal Antibody
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542687
Description
Peptide sequence: TYSVKEMEYL TRAIDRTGGE KNTVIVISLG QHFRPFPIDV FIRRALNVHK Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
The function remains unknown.
            Specifications
| FAM55D | |
| Polyclonal | |
| Unconjugated | |
| NXPE4 | |
| C11orf33; C130036J11; D930028F11Rik; Fam55b; Fam55d; family with sequence similarity 55, member B; family with sequence similarity 55, member D; hypothetical protein LOC244853; hypothetical protein LOC515648; LOC100731209; neurexophilin and PC-esterase domain family member 4; neurexophilin and PC-esterase domain family, member 4; NXPE family member 4; Nxpe4; Protein FAM55D; UNQ3018/PRO9799 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 54827 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | 
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q6UWF7 | |
| NXPE4 | |
| Synthetic peptide directed towards the C-terminal of human FAM55D (aa 407-456). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG | 
Safety and Handling
                                                    
                                                    WARNING: Cancer - www.P65Warnings.ca.gov 
                                                
                                            
                                        
                                    
                                
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction