Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM55D Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560128
Description
Immunogen sequence: LSKQEKSLFE RSNVGVEIME KFNTISVSKC NKETVAMKEK CKFGMTSTIP SGHVWRNTWN PVSCSLATVK MKE Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 37%.
The function remains unknown.
Specifications
FAM55D | |
Polyclonal | |
Unconjugated | |
NXPE4 | |
C11orf33; C130036J11; D930028F11Rik; Fam55b; Fam55d; family with sequence similarity 55, member B; family with sequence similarity 55, member D; hypothetical protein LOC244853; hypothetical protein LOC515648; LOC100731209; neurexophilin and PC-esterase domain family member 4; neurexophilin and PC-esterase domain family, member 4; NXPE family member 4; Nxpe4; Protein FAM55D; UNQ3018/PRO9799 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54827 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6UWF7 | |
NXPE4 | |
Recombinant protein corresponding to Human FAM55D. Recombinant protein control fragment (Product #RP-97345). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction