Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM55D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5107202
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5107202 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5107202 Supplier Invitrogen™ Supplier No. PA5107202
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sequence homology: Cow: 92%; Dog: 85%; Guinea Pig: 92%; Horse: 92%; Human: 100%; Mouse: 85%; Pig: 93%; Rabbit: 93%; Rat: 92%.

The function remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM55D
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene NXPE4
Gene Accession No. Q52KP5
Gene Alias C11orf33; C130036J11; D930028F11Rik; Fam55b; Fam55d; family with sequence similarity 55, member B; family with sequence similarity 55, member D; hypothetical protein LOC244853; hypothetical protein LOC515648; LOC100731209; neurexophilin and PC-esterase domain family member 4; neurexophilin and PC-esterase domain family, member 4; NXPE family member 4; Nxpe4; Protein FAM55D; UNQ3018/PRO9799
Gene Symbols NXPE4
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal region of Mouse Fam55d.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 244853
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.