Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM55D Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS8196R

Description
The function remains unknown.Specifications
FAM55D | |
Polyclonal | |
Unconjugated | |
NXPE4 | |
BOS_14363; C11orf33; C130036J11; D930028F11Rik; Fam55b; Fam55d; family with sequence similarity 55, member B; family with sequence similarity 55, member D; hypothetical protein LOC244853; LOC100731209; neurexophilin and PC-esterase domain family member 4; neurexophilin and PC-esterase domain family, member 4; NXPE family member 4; Nxpe4; Protein FAM55D; UNQ3018/PRO9799 | |
Rabbit | |
Protein A | |
RUO | |
54827 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q6UWF7 | |
NXPE4 | |
KLH conjugated synthetic peptide derived from human FAM55D. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction