Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM57A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571453
Description
FAM57A Polyclonal Antibody for Western Blot
The protein encoded by this gene is a membrane-associated protein that promotes lung carcinogenesis. The encoded protein may be involved in amino acid transport and glutathione metabolism since it can interact with a solute carrier family member (SLC3A2) and an isoform of gamma-glutamyltranspeptidase-like 3. An alternatively spliced variant encoding a protein that lacks a 32 aa internal segment showed the opposite effect, inhibiting lung cancer cell growth. Knockdown of this gene also inhibited lung carcinogenesis and tumor cell growth. Several transcript variants encoding different isoforms have been found for this gene.
Specifications
| FAM57A | |
| Polyclonal | |
| Unconjugated | |
| FAM57A | |
| CT120; fa57a; FAM57A; family with sequence similarity 57 member A; family with sequence similarity 57, member A; membrane protein expressed in epithelial-like lung adenocarcinoma; Protein CT120; Protein FAM57A; TLC domain-containing protein 3A; TLCD3A | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 79850 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8TBR7 | |
| FAM57A | |
| synthetic peptide directed towards the C terminal region of human FAM57A. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction