Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM57B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569656
Description
This target displays homology in the following species: Cow: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 86%; Rat: 100%.
FAM57B is a protein coding gene.
Specifications
FAM57B | |
Polyclonal | |
Unconjugated | |
FAM57B | |
1500016O10Rik; A330104J06Rik; AI413816; AW060769; Ceramide synthase; ceramide synthase; protein FAM57B; FAM57B; family with sequence similarity 57 member B; family with sequence similarity 57, member B; FP1188; Protein FAM57B; RGD1308215; TLC domain ceramide synthase 3B; TLC domain containing 3B; TLC domain-containing protein 3B; TLCD3B | |
Rabbit | |
Affinity chromatography | |
RUO | |
83723 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q71RH2 | |
FAM57B | |
synthetic peptide directed towards the C-terminal region of human FAM57B. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction