Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM58A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566284
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566284 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566284 Supplier Invitrogen™ Supplier No. PA566284
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FRVARFIMEAG VKLGMRSIPI ATACTIYHKF FCETNLDAYD PYLIAMSSIY LAGKVEEQHL RTRDIINVSN RYFNP Highest antigen sequence identity to the following orthologs - mouse 92%, rat 91%.

Activating cyclin for the cyclin-associated kinase CDK10.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM58A
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCNQ
Gene Accession No. Q8N1B3
Gene Alias Ccnq; CDK10-activating cyclin; cyclin M; cyclin Q; cyclin-M; Cyclin-Q; Cyclin-related protein FAM58A; Fam58a; Fam58b; family with sequence similarity 58 member A; family with sequence similarity 58, member A; family with sequence similarity 58, member B; RGD1305651; STAR
Gene Symbols CCNQ
Host Species Rabbit
Immunogen Recombinant Human FAM58A. Recombinant protein control fragment (Product #RP-106969).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92002
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.