Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM58A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100729
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100729 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100729 Supplier Invitrogen™ Supplier No. PA5100729
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total FA58A.

Activating cyclin for the cyclin-associated kinase CDK10.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM58A
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CCNQ
Gene Accession No. Q4QQW5, Q8N1B3
Gene Alias Ccnq; CDK10-activating cyclin; cyclin M; cyclin Q; cyclin-M; Cyclin-Q; Cyclin-related protein FAM58A; Fam58a; Fam58b; family with sequence similarity 58 member A; family with sequence similarity 58, member A; family with sequence similarity 58, member B; RGD1305651; STAR
Gene Symbols CCNQ
Host Species Rabbit
Immunogen A synthesized peptide derived from human FAM58A(Accession Q8N1B3), corresponding to amino acid residues C60-L110.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 303321, 92002
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.