Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM59A Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281933MFV500
Description
FAM59A Polyclonal antibody specifically detects FAM59A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| FAM59A | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| Chromosome 18 Open Reading Frame 11, chromosome 18 open reading frame 11, family with sequence, FAM59A, Family With Sequence Similarity 59, Member A, FLJ21610, GAREM, Gm944, GRB2 associated, regulator of MAPK1, Grb2-Associated And Regulator Of Erk/MAPK, GRB2-Associated And Regulator Of MAPK Protein, GRB2-Associated And Regulator Of MAPK1, Protein FAM59A | |
| FAM59A antibody was raised against a 19 amino acid synthetic peptide from near the carboxy terminus of human FAM59A. The immunogen is located within amino acids 630 - 680 of FAM59A. Amino Acid Squence: DPSRSTSTPRQKTPGTPKR | |
| 0.1 mL | |
| Cancer | |
| 64762 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction