Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM59A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP247460
Description
FAM59A Polyclonal specifically detects FAM59A in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
FAM59A | |
Polyclonal | |
Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Chromosome 18 Open Reading Frame 11, chromosome 18 open reading frame 11, family with sequence, FAM59A, Family With Sequence Similarity 59, Member A, FLJ21610, GAREM, Gm944, GRB2 associated, regulator of MAPK1, Grb2-Associated And Regulator Of Erk/MAPK, GRB2-Associated And Regulator Of MAPK Protein, GRB2-Associated And Regulator Of MAPK1, Protein FAM59A | |
Rabbit | |
Affinity Purified | |
RUO | |
64762 | |
Human | |
IgG |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
GAREM1 | |
This antibody was developed against a recombinant protein corresponding to amino acids: ILPMHFPLHLTVPKFSLPEHLVKGESWPETLVHHWLGICQEQFDIDEYSRAVRDVKTDWNEECKSPKKGRCSGHNHVPNSLSYARDELTQS | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction