Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM59A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559260
Description
Immunogen sequence: ILPMHFPLHL TVPKFSLPEH LVKGESWPET LVHHWLGICQ EQFDIDEYSR AVRDVKTDWN EECKSPKKGR CSGHNHVPNS LSYARDELTQ S Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 35%.
FAM59A is a recently identified intracellular protein belonging to the FAM59 family. It contains one SAM (sterile alpha motif) domain. Alternative splicing result shows that it has three isoforms. The function of FAM59A is yet unknown. It is widely expressed in many tissues.
Specifications
FAM59A | |
Polyclonal | |
Unconjugated | |
GAREM1 | |
C18orf11; C86169; Fam59a; family with sequence similarity 59, member A; GAREM; GAREM1; Gm944; GRB2 associated regulator of MAPK1 1; GRB2 associated regulator of MAPK1 subtype 1; GRB2 associated, regulator of MAPK1; Grb2-associated and regulator of Erk/MAPK; GRB2-associated and regulator of MAPK protein; GRB2-associated and regulator of MAPK protein 1; GRB2-associated and regulator of MAPK1; Kiaa4238; mKIAA4238; protein FAM59A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
64762 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H706 | |
GAREM1 | |
Recombinant protein corresponding to Human FAM59A. Recombinant protein control fragment (Product #RP-98276). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction