Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM59A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559841
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559841 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559841 Supplier Invitrogen™ Supplier No. PA559841
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GNSEVNLHGC RDLGGDWAPF PHDILPYQDS GDSGSDYLFP EASEESAGIP GKSELPYEEL WLEEGKPSHQ PLTRSLSEKN RCDQFRGSVR SKCATSPLPI P Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 24%.

FAM59A is a recently identified intracellular protein belonging to the FAM59 family. It contains one SAM (sterile alpha motif) domain. Alternative splicing result shows that it has three isoforms. The function of FAM59A is yet unknown. It is widely expressed in many tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM59A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GAREM1
Gene Accession No. Q9H706
Gene Alias C18orf11; C86169; Fam59a; family with sequence similarity 59, member A; GAREM; GAREM1; Gm944; GRB2 associated regulator of MAPK1 1; GRB2 associated regulator of MAPK1 subtype 1; GRB2 associated, regulator of MAPK1; Grb2-associated and regulator of Erk/MAPK; GRB2-associated and regulator of MAPK protein; GRB2-associated and regulator of MAPK protein 1; GRB2-associated and regulator of MAPK1; Kiaa4238; mKIAA4238; protein FAM59A
Gene Symbols GAREM1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM59A. Recombinant protein control fragment (Product #RP-98275).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64762
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.