Learn More
Invitrogen™ FAM59B Polyclonal Antibody

Description
A suggested positive control is SK-N-SH cell lysate. PA5-20846 can be used with blocking peptide PEP-0960.
Specifications
Specifications
| Antigen | FAM59B |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | GAREM2 |
| Gene Accession No. | Q6PAJ3, Q75VX8 |
| Gene Alias | FAM59B; family with sequence similarity 59, member B; Garem2; Gareml; Gm444; GRB2 associated regulator of MAPK1 2; GRB2 associated regulator of MAPK1 subtype 2; GRB2 associated, regulator of MAPK1-like; GRB2-associated and regulator of MAPK protein 2; GRB2-associated and regulator of MAPK protein-like; GRB2-associated and regulator of MAPK1-like; HRIHFB2063; KIAA2038; protein FAM59B |
| Gene Symbols | GAREM2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.