Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM59B Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988372
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8372 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8372 Supplier Bioss Supplier No. BS8217R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM59B is related to the FAM59A protein (also known as GAREM). While little is known of the role of the FAM59B protein, FAM59A is a downstream molecule in the EGF signaling pathway and binds Grb2, an adaptor protein crucial to the transduction of growth signals from the plasma membrane to the nucleus. Like FAM59A, FAM59B may play a role in ligand-mediated signaling pathway of the EGF receptor and the tumorigenesis of cells.

Specifications

Antigen FAM59B
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GAREM2
Gene Accession No. Q75VX8
Gene Alias Fam59b; family with sequence similarity 59, member B; GAREM2; Gareml; Gm444; GRB2 associated regulator of MAPK1 2; GRB2 associated regulator of MAPK1 subtype 2; GRB2 associated, regulator of MAPK1-like; GRB2-associated and regulator of MAPK protein 2; GRB2-associated and regulator of MAPK protein-like; GRB2-associated and regulator of MAPK1-like; HRIHFB2063; KIAA2038; protein FAM59B
Gene Symbols GAREM2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from hu FAM59B.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150946
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.