Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM5C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563007
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563007 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563007 Supplier Invitrogen™ Supplier No. PA563007
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FLYCNENGLL GSFSEETHSC TCPNDQVVCT AFLPCTVGDA SACLTCAPDN RTR Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

Inhibits neuronal cell proliferation by negative regulation of the cell cycle transition. Promotes pituitary gonadotrope cell proliferation, migration and invasion, when overexpressed. May play a role in cell pituitary tumor development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM5C
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BRINP3
Gene Accession No. Q76B58
Gene Alias B830045N13Rik; BMP/retinoic acid inducible neural specific 3; BMP/retinoic acid-inducible neural-specific protein 3; bone morphogenetic protein/retinoic acid inducible neural specific 3; bone morphogenetic protein/retinoic acid inducible neural-specific 3; BRINP3; DBCCR1L; DBCCR1L1; DBCCR1-like protein 1; FAM5C; family with sequence similarity 5, member C; protein FAM5C; RP11-445K1.1
Gene Symbols BRINP3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM5C. Recombinant protein control fragment (Product #RP-99804).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 339479
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.