Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM5C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598528
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598528 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598528 Supplier Invitrogen™ Supplier No. PA598528
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM5C Polyclonal Antibody for Western Blot, IHC (P), ELISA

Inhibits neuronal cell proliferation by negative regulation of the cell cycle transition. Promotes pituitary gonadotrope cell proliferation, migration and invasion, when overexpressed. May play a role in cell pituitary tumor development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM5C
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3.78 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene BRINP3
Gene Accession No. Q76B58
Gene Alias B830045N13Rik; BMP/retinoic acid inducible neural specific 3; BMP/retinoic acid-inducible neural-specific protein 3; bone morphogenetic protein/retinoic acid inducible neural specific 3; bone morphogenetic protein/retinoic acid inducible neural-specific 3; BRINP3; DBCCR1L; DBCCR1L1; DBCCR1-like protein 1; FAM5C; family with sequence similarity 5, member C; protein FAM5C; RP11-445K1.1
Gene Symbols BRINP3
Host Species Rabbit
Immunogen Recombinant Human BMP/retinoic acid-inducible neural-specific protein 3 protein (389-487AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 339479
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.