Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM5C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116811
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116811 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116811 Supplier Invitrogen™ Supplier No. PA5116811
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: U-251MG, Mouse brain Immunogen sequence: LQGLQVLLPD YLQERFVQAA LSYIACNSEG EFICKENDCW CHCGPKFPEC NCPSMDIQAM EENLLRITET WKAYNSDFEE SDEFKLFMKR LPMNYFLNTS TIMHLWTMDS NFQRRYEQLE NSMKQLFLKA QKIVHKLFSL SKRCHKQPLI SLPRQRTSTY WLTRIQSFLY CNENGLLGSF SEETHSCTCP NDQVVCTAFL PCTVGDASAC LTCAPDNRTR CGTCNTGYML SQGLCKPEVA ESTDHYIGFE TDLQDLEMKY L.

Inhibits neuronal cell proliferation by negative regulation of the cell cycle transition. Promotes pituitary gonadotrope cell proliferation, migration and invasion, when overexpressed. May play a role in cell pituitary tumor development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM5C
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.19 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene BRINP3
Gene Accession No. Q499E0, Q76B58, Q8K1M7
Gene Alias B830045N13Rik; BMP/retinoic acid inducible neural specific 3; BMP/retinoic acid-inducible neural-specific protein 3; bone morphogenetic protein/retinoic acid inducible neural specific 3; bone morphogenetic protein/retinoic acid inducible neural-specific 3; BRINP3; DBCCR1L; DBCCR1L1; DBCCR1-like protein 1; FAM5C; family with sequence similarity 5, member C; protein FAM5C; RP11-445K1.1
Gene Symbols BRINP3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 240-500 of human BRINP3 (NP_9502521).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 215378, 286901, 339479
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.