Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM60A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584353
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584353 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584353 Supplier Invitrogen™ Supplier No. PA584353
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RAKSSSSRFT DSKRYEKDFQ SCFGLHETRS GDICNACVLL VKRWKKLPAG SKKNWNHVVD ARAGPSLKTT LKPKKVKTLS GN Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.

FAM60A is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM60A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SINHCAF
Gene Accession No. Q9NP50
Gene Alias C12orf14; FAM60A; family with sequence similarity 60 member A; family with sequence similarity 60, member A; L4; Protein FAM60A; SIN3-HDAC complex-associated factor; SINHCAF; TERA; Tera protein homolog
Gene Symbols SINHCAF
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SINHCAF. Recombinant protein control fragment (Product #RP-108425).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58516
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.