Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM62A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111742
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111742 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111742 Supplier Invitrogen™ Supplier No. PA5111742
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSGPNSRLYM KLVMRILYLD SSEICFPTVP GCPGAWDVDS ENPQRGSSVD APPRPCHTTP DSQFGTEHVL RIHVLEAQDL IAKDRF Highest antigen sequence indentity to the following orthologs: Rat - 81%, Mouse - 81%.

May play a role as calcium-regulated intrinsic membrane protein. Target protein is an enriched in adipocyte plasma membranes and high density microsome fractions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM62A
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ESYT1
Gene Accession No. Q9BSJ8
Gene Alias ESYT1; E-Syt1; extended synaptotagmin 1; extended synaptotagmin like protein 1; extended synaptotagmin protein 1; extended synaptotagmin-1; extended synaptotagmin-like protein 1; FAM62A; family with sequence similarity 62 (C2 domain containing), member A; KIAA0747; MBC2; membrane bound C2 domain containing protein; membrane-bound C2 domain-containing protein; vp115
Gene Symbols ESYT1
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human FAM62A. Recombinant protein control fragment (Product #RP-108102).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23344
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.