Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM62B Monoclonal Antibody (C4-D0)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA542563
This item is not returnable.
View return policy
Description
Positive Control: Siha cell lysate, THP-1 cell lysate, human colon carcinoma tissue. Subcellular Location: Cell membrane, Endoplasmic reticulum, Membrane.
Tethers the endoplasmic reticulum to the cell membrane and promotes the formation of appositions between the endoplasmic reticulum and the cell membrane. Binds glycerophospholipids in a barrel-like domain and may play a role in cellular lipid transport. Plays a role in FGF signaling via its role in the rapid internalization of FGFR1 that has been activated by FGF1 binding; this occurs most likely via the AP-2 complex. Promotes the localization of SACM1L at endoplasmic reticulum-plasma membrane contact sites (EPCS) (PubMed:27044890). [UniProt].
Specifications
FAM62B | |
Monoclonal | |
2 mg/mL | |
TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
A0FGR8 | |
ESYT2 | |
Recombinant protein within human FAM62B aa 300-550. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
C4-D0 | |
Unconjugated | |
ESYT2 | |
2410017M09Rik; 4921504I16Rik; chr2 synaptotagmin; CHR2SYT; D12Ertd551e; ESYT2; E-Syt2; extended synaptotagmin 2; extended synaptotagmin like protein 2; extended synaptotagmin protein 2; extended synaptotagmin-2; extended synaptotagmin-like protein 2; Fam62b; family with sequence similarity 62 (C2 domain containing), member B; family with sequence similarity 62, member B; KIAA1228; RGD1565705 | |
Mouse | |
Protein G | |
RUO | |
57488 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction