Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM62B Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144991
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144991 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144991 Supplier Invitrogen™ Supplier No. PA5144991
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DHQHSAQVKR PSVSKEGRKT SIKSHMSGSP GPGGSNTAPS TPVIGGSDKP GMEEKAQPPE AGPQGLHDQG RSSS Antigen sequence identity: Mouse 77%; Rat 77%.

Tethers the endoplasmic reticulum to the cell membrane and promotes the formation of appositions between the endoplasmic reticulum and the cell membrane. Binds glycerophospholipids in a barrel-like domain and may play a role in cellular lipid transport. Plays a role in FGF signaling via its role in the rapid internalization of FGFR1 that has been activated by FGF1 binding; this occurs most likely via the AP-2 complex. Promotes the localization of SACM1L at endoplasmic reticulum-plasma membrane contact sites (EPCS) (PubMed:27044890). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM62B
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ESYT2
Gene Accession No. A0FGR8
Gene Alias 2410017M09Rik; 4921504I16Rik; chr2 synaptotagmin; CHR2SYT; D12Ertd551e; ESYT2; E-Syt2; extended synaptotagmin 2; extended synaptotagmin like protein 2; extended synaptotagmin protein 2; extended synaptotagmin-2; extended synaptotagmin-like protein 2; Fam62b; family with sequence similarity 62 (C2 domain containing), member B; family with sequence similarity 62, member B; KIAA1228; RGD1565705
Gene Symbols ESYT2
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 665-738. Recombinant protein control fragment (Product #RP-109877).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57488
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.