Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM62C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558693
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558693 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558693 Supplier Invitrogen™ Supplier No. PA558693
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PYADLTLEQR FQLDHSGLDS LISMRLVLRF LQVEERELGS PYTGPEALKK GPLLIKKVAT NQGPKAQPQE EGPTDLPCPP DPASDTKD Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 72%.

FAM62C belongs to the extended synaptotagmin family. It is a single-pass membrane protein, and contains 3 C2 domains. FAM62C may play a role as calcium-regulated intrinsic membrane protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM62C
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ESYT3
Gene Accession No. A0FGR9
Gene Alias chr3 synaptotagmin; CHR3SYT; D930024E11; D9Ertd280e; Esyt3; E-Syt3; extended synaptotagmin 3; extended synaptotagmin protein 3; extended synaptotagmin-3; extended synaptotagmin-like protein 3; Fam62c; family with sequence similarity 62 (C2 domain containing), member C; family with sequence similarity 62, member C; Kiaa4186; mKIAA4186; RGD1561304
Gene Symbols ESYT3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM62C. Recombinant protein control fragment (Product #RP-96573).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83850
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.