Learn More
Invitrogen™ FAM62C Polyclonal Antibody

Description
Sequence homology: Cow: 86%; Dog: 93%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 86%; Rat: 100%.
Specifications
Specifications
| Antigen | FAM62C |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 |
| Gene | ESYT3 |
| Gene Accession No. | A0FGR9 |
| Gene Alias | chr3 synaptotagmin; CHR3SYT; D930024E11; D9Ertd280e; Esyt3; E-Syt3; extended synaptotagmin 3; extended synaptotagmin protein 3; extended synaptotagmin-3; extended synaptotagmin-like protein 3; Fam62c; family with sequence similarity 62 (C2 domain containing), member C; family with sequence similarity 62, member C; Kiaa4186; mKIAA4186; RGD1561304 |
| Gene Symbols | ESYT3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.