Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM63A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566362
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566362 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566362 Supplier Invitrogen™ Supplier No. PA566362
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YLIALSLQQQQ PRGPLGLTDL ELAQQLQQEE YQQQQAAQPV RMRTRVLSLQ GRGATSGRPA GERRQRPKHE SDCI Highest antigen sequence identity to the following orthologs - mouse 83%, rat 82%.

Hydrolase that can specifically remove 'Lys-48'-linked conjugated ubiquitin from proteins. Has exodeubiquitinase activity and has a preference for long polyubiquitin chains. May play a regulatory role at the level of protein turnover.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM63A
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MINDY1
Gene Accession No. Q8N5J2
Gene Alias 1810005H09Rik; 4930504E06Rik; AA562754; AA960384; AW561904; cI-40; Deubiquitinating enzyme MINDY-1; FAM63A; family with sequence similarity 63 member A; family with sequence similarity 63, member A; KIAA1390; Mindy1; mKIAA1390; NF-E2 inducible protein; Ni; Protein FAM63A; RP11-316M1.5; Ubiquitin carboxyl-terminal hydrolase MINDY-1
Gene Symbols MINDY1
Host Species Rabbit
Immunogen Recombinant Human FAM63A. Recombinant protein control fragment (Product #RP-105167).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55793
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.