Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM63A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113220
Description
FAM63A Polyclonal Antibody for Western Blot, ELISA
Hydrolase that can specifically remove 'Lys-48'-linked conjugated ubiquitin from proteins. Has exodeubiquitinase activity and has a preference for long polyubiquitin chains. May play a regulatory role at the level of protein turnover.
Specifications
| FAM63A | |
| Polyclonal | |
| Unconjugated | |
| MINDY1 | |
| 1810005H09Rik; 4930504E06Rik; AA562754; AA960384; AW561904; cI-40; Deubiquitinating enzyme MINDY-1; FAM63A; family with sequence similarity 63 member A; family with sequence similarity 63, member A; KIAA1390; Mindy1; mKIAA1390; NF-E2 inducible protein; Ni; Protein FAM63A; RP11-316M1.5; Ubiquitin carboxyl-terminal hydrolase MINDY-1 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 55793, 75007 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q76LS9, Q8N5J2 | |
| MINDY1 | |
| Recombinant Human Ubiquitin carboxyl-terminal hydrolase MINDY-1 protein (3-278AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction