Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM63B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566614
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566614 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566614 Supplier Invitrogen™ Supplier No. PA566614
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSSAGLDLKDS GLESPAAAEA PLRGQYKVTA SPETAVAGVG HELGTAGDAG ARPDLAGTCQ AELTA Highest antigen sequence identity to the following orthologs - mouse 50%, rat 52%.

Hydrolase that can remove 'Lys-48'-linked conjugated ubiquitin from proteins (PubMed:27292798). Binds to polyubiquitin chains of different linkage types, including 'Lys-6', 'Lys-11', 'Lys-29', 'Lys-33', 'Lys-48' and 'Lys-63' (PubMed:28082312). May play a regulatory role at the level of protein turnover (PubMed:27292798). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM63B
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MINDY2
Gene Accession No. Q8NBR6
Gene Alias Deubiquitinating enzyme MINDY-2; FAM63B; family with sequence similarity 63 member B; family with sequence similarity 63, member B; KIAA1164; MINDY2; Protein FAM63B; Ubiquitin carboxyl-terminal hydrolase MINDY-2
Gene Symbols MINDY2
Host Species Rabbit
Immunogen Recombinant Human FAM63B. Recombinant protein control fragment (Product #RP-107273).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54629
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.