Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM63B Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA562318

Catalog No. PIPA562318


Only null left
Add to Cart

Description

Description

Immunogen sequence: SPPGESPSLD SLESFSNLHS FPSSCEFNSE EGAENRVPEE EEGAAVLPGA VPLCKEEEGE ETAQVLAASK ERFPGQSVYH IKWIQWKE Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 85%.

Hydrolase that can remove 'Lys-48'-linked conjugated ubiquitin from proteins (PubMed:27292798). Binds to polyubiquitin chains of different linkage types, including 'Lys-6', 'Lys-11', 'Lys-29', 'Lys-33', 'Lys-48' and 'Lys-63' (PubMed:28082312). May play a regulatory role at the level of protein turnover (PubMed:27292798). [UniProt].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FAM63B
Polyclonal
Unconjugated
MINDY2
Deubiquitinating enzyme MINDY-2; FAM63B; family with sequence similarity 63 member B; family with sequence similarity 63, member B; KIAA1164; MINDY2; Protein FAM63B; Ubiquitin carboxyl-terminal hydrolase MINDY-2
Rabbit
Antigen affinity chromatography
RUO
54629
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8NBR6
MINDY2
Recombinant protein corresponding to Human FAM63B. Recombinant protein control fragment (Product #RP-101257).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.