Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM65B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104330
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104330 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104330 Supplier Invitrogen™ Supplier No. PA5104330
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total FAM65B.

Required for hearing (PubMed:24958875). Involved in skeletal muscle development (PubMed:24687993).
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM65B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Ripor2
Gene Accession No. Q7TP54, Q80U16, Q9Y4F9
Gene Alias Ab2-162; C6orf32; DFNB104; DIFF40; DIFF48; FAM65B; family with sequence similarity 65 member B; family with sequence similarity 65, member B; Kiaa0386; myogenesis-related and NCAM-associated protein homolog; MYONAP; PL48; protein FAM65B; RGD1306939; Rho family-interacting cell polarization regulator 2; RIPOR2
Gene Symbols Ripor2
Host Species Rabbit
Immunogen A synthesized peptide derived from human RIPOR2(Accession Q9Y4F9), corresponding to amino acid residues E312-L362.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 193385, 306934, 9750
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.