Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM65B Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS12370R

Description
Required for hearing (PubMed:24958875). Involved in skeletal muscle development (PubMed:24687993).Specifications
FAM65B | |
Polyclonal | |
Unconjugated | |
Ripor2 | |
Ab2-162; C6orf32; DFNB104; DIFF40; DIFF48; FAM65B; family with sequence similarity 65 member B; family with sequence similarity 65, member B; KIAA0386; myogenesis-related and NCAM-associated protein homolog; MYONAP; PL48; protein FAM65B; RGD1306939; Rho family-interacting cell polarization regulator 2; RIPOR2 | |
Rabbit | |
Protein A | |
RUO | |
9750 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9Y4F9 | |
Ripor2 | |
KLH conjugated synthetic peptide derived from human FAM65B. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction